Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dync1h1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019226.3 | NP_062099.3 | 40 | 8186 | ||
GenBank mRNA | L08505.1 | - | 40 | 8186 |