Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Erbb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017218.3 | NP_058914.2 | 8 | 1076 | ||
GenBank mRNA | U29339.2 | - | 8 | 1076 |