Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc12a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019229.2 | NP_062102.1 | 22 | 3018 | ||
XM_006255482.2 | XP_006255544.1 | 22 | 4422 | |||
XM_008772492.1 | XP_008770714.1 | 21 | 3009 | |||
GenBank mRNA | FQ219935.1 | - | 4 | 574 | ||
U55815.1 | - | 22 | 3018 |