Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cox7a2l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751367.3 | XP_003751415.1 | 1 | 94 |
Gene Symbol: Cox7a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022503.2 | NP_071948.1 | 2 | 143 | ||
GenBank mRNA | BC166880.1 | - | 2 | 111 | 6888806 | |
BG668415.1 | - | 2 | 150 | |||
FQ209801.1 | - | 2 | 138 | |||
FQ209847.1 | - | 2 | 110 | |||
FQ210551.1 | - | 2 | 110 | |||
FQ211276.1 | - | 2 | 111 | |||
FQ214317.1 | - | 2 | 119 | |||
FQ217361.1 | - | 2 | 110 | |||
FQ221839.1 | - | 2 | 115 | |||
FQ223968.1 | - | 2 | 134 | |||
FQ228537.1 | - | 2 | 182 | |||
FQ229029.1 | - | 2 | 112 | |||
FQ229035.1 | - | 2 | 144 | |||
FQ230547.1 | - | 2 | 171 | |||
X54080.1 | - | 2 | 117 |