Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | U31777.1 | - | 8 | 3561 |
Gene Symbol: LOC100911672 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750658.3 | XP_003750706.1 | 8 | 3551 | ||
XM_006244637.2 | XP_006244699.1 | 8 | 3537 |