Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Padi3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017230.2 | NP_058926.1 | 14 | 1663 | ||
GenBank mRNA | D88034.1 | - | 14 | 1664 |