Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amhr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030998.1 | NP_112260.1 | 2 | 262 | ||
GenBank mRNA | KJ923229.1 | - | 2 | 262 | ||
U42427.1 | - | 2 | 262 | |||
X71916.1 | - | 2 | 244 |