Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccl20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019233.1 | NP_062106.1 | 2 | 166 | ||
XM_006245164.2 | XP_006245226.1 | 2 | 232 | |||
GenBank mRNA | AF053312.1 | - | 2 | 167 | ||
U90447.1 | - | 2 | 166 |