Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lcn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024136.1 | NP_077050.1 | 1, 2 | 136 | ||
XM_008761580.1 | XP_008759802.1 | 1, 2 | 1201 | |||
GenBank mRNA | M12790.1 | - | 1, 2 | 88 | ||
X59832.1 | - | 1, 2 | 128 |