Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMELX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142.2 | NP_001133.1 | 5 | 397 | ||
NM_182680.1 | NP_872621.1 | 6 | 439 | |||
NM_182681.1 | NP_872622.1 | 4 | 349 | |||
XM_017029404.1 | XP_016884893.1 | 4 | 1498 | |||
GenBank mRNA | AF436849.1 | - | 6 | 439 | ||
BC069118.1 | - | 5 | 397 | 7216972 | ||
BC074951.2 | - | 5 | 376 | 30915277 | ||
M86932.1 | - | 5 | 397 | |||
S67147.1 | - | 1 | 185 |