Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lsamp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017242.1 | NP_058938.1 | 1 | 153 | ||
XM_006248339.2 | XP_006248401.1 | 1 | 604 | |||
GenBank mRNA | AY326256.1 | - | 1 | 122 | ||
BC087607.1 | - | 1 | 124 | 7314244 | ||
U31554.1 | - | 1 | 153 |