Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUMO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006936.2 | NP_008867.2 | 2, 3 | 314 | ||
GenBank mRNA | AK222591.1 | - | 2, 3 | 245 | ||
AK312350.1 | - | 2, 3 | 245 | |||
BC000036.2 | - | 2, 3 | 239 | 3505840 | ||
BC008420.1 | - | 2, 3 | 314 | 4091864 | ||
BT007008.1 | - | 2, 3 | 153 | |||
CR542173.1 | - | 2, 3 | 153 | |||
CR542188.1 | - | 2, 3 | 153 | |||
X99584.1 | - | 2, 3 | 247 |