Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Htr2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017250.1 | NP_058946.1 | 3 | 1572 | ||
XM_008767216.1 | XP_008765438.1 | 4 | 2422 | |||
GenBank mRNA | X66842.1 | - | 3 | 1572 |
Set Membership: |
G-protein coupled receptor |