Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Htr2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017250.1 | NP_058946.1 | 1 | 542 | ||
XM_006245369.2 | XP_006245431.1 | 1 | 2245 | |||
XM_008767216.1 | XP_008765438.1 | 2 | 1392 | |||
GenBank mRNA | X66842.1 | - | 1 | 542 |
Set Membership: |
G-protein coupled receptor |