Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gjb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017251.2 | NP_058947.1 | 2 | 806 | ||
XM_006257061.2 | XP_006257123.1 | 3 | 958 | |||
XM_008773255.1 | XP_008771477.1 | 2 | 1689 | |||
GenBank mRNA | BC078868.1 | - | 2 | 770 | 7127952 | |
FQ210131.1 | - | 2 | 791 | |||
X04070.1 | - | 1 | 752 |