Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUMO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286416.1 | NP_001273345.1 | 4 | 528 | ||
NM_006936.2 | NP_008867.2 | 4 | 414 | |||
GenBank mRNA | AK222591.1 | - | 4 | 345 | ||
AK300822.1 | - | 4 | 480 | |||
AK312350.1 | - | 4 | 345 | |||
BC000036.2 | - | 4 | 339 | 3505840 | ||
BC008420.1 | - | 4 | 414 | 4091864 | ||
BT007008.1 | - | 4 | 253 | |||
CR542173.1 | - | 4 | 253 | |||
CR542188.1 | - | 4 | 253 | |||
X99584.1 | - | 4 | 347 |