Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053311.1 | NP_445763.1 | 10 | 1832 | ||
XM_006241294.2 | XP_006241356.1 | 10 | 1924 | |||
XM_006241295.2 | XP_006241357.1 | 9 | 1801 | |||
XM_006241296.2 | XP_006241358.1 | 10 | 1924 | |||
GenBank mRNA | J03753.1 | - | 10 | 1832 |