Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUMO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005849.1 | NP_001005849.1 | 3 | 890 | ||
NM_006937.3 | NP_008868.3 | 4 | 962 | |||
GenBank mRNA | AK024823.1 | - | 4 | 863 | ||
AK225090.1 | - | 4 | 875 | |||
BC008450.1 | - | 4 | 929 | 4133768 | ||
BC016775.1 | - | 4 | 943 | 4096440 | ||
BC022340.1 | - | 4 | 913 | 4134531 | ||
BC062713.1 | - | 4 | 876 | 30404344 | ||
BC068465.1 | - | 4 | 928 | 5277638 | ||
BC070159.1 | - | 4 | 936 | 6592382 | ||
BC071645.1 | - | 4 | 869 | 3884132 | ||
BC071646.1 | - | 4 | 907 | 4369354 | ||
BC107853.1 | - | 4 | 868 | 5772748 |