Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alox5ap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017260.2 | NP_058956.1 | 1 | 106 | ||
GenBank mRNA | BC086593.1 | - | 1 | 106 | 7310271 | |
X52196.1 | - | 1 | 88 |