Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMELY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143.1 | NP_001134.1 | 4, 5 | 207 | ||
XM_017030042.1 | XP_016885531.1 | 5, 6 | 278 | |||
GenBank mRNA | AY487421.1 | - | 4, 5 | 181 | ||
BC069138.1 | - | 4, 5 | 207 | 7216997 | ||
BC074976.1 | - | 4, 5 | 168 | 30915513 | ||
BC074977.1 | - | 4, 5 | 168 | 30915296 | ||
M86933.1 | - | 4, 5 | 207 |