Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arntl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024362.2 | NP_077338.2 | 13 | 1270 | ||
XM_006230029.2 | XP_006230091.1 | 11 | 1028 | |||
XM_006230030.2 | XP_006230092.1 | 11 | 1028 | |||
XM_008759716.1 | XP_008757938.1 | 11 | 968 | |||
GenBank mRNA | AB012600.1 | - | 8 | 702 | ||
AF015953.1 | - | 7 | 599 | |||
AF317669.2 | - | 8 | 600 | |||
BC099833.1 | - | 13 | 1270 | 7936079 |