Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp2a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110139.2 | NP_001103609.1 | 12 | 1965 | ||
NM_001110823.2 | NP_001104293.1 | 12 | 1965 | |||
GenBank mRNA | AF043106.1 | - | 7 | 937 | ||
J04022.1 | - | 12 | 1963 | |||
J04023.1 | - | 12 | 1598 | |||
J04024.1 | - | 12 | 1697 | |||
X15635.1 | - | 12 | 1666 |