Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bckdhb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019267.1 | NP_062140.1 | 8 | 916 | ||
XM_006243450.2 | XP_006243512.1 | 9 | 973 | |||
GenBank mRNA | BC158861.1 | - | 8 | 909 | 7932093 | |
CB547976.1 | - | 4 | 337 | |||
FQ213896.1 | - | 8 | 898 | |||
FQ226240.1 | - | 8 | 904 | |||
M94040.1 | - | 8 | 827 |