Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc8a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270772.1 | NP_001257701.1 | 2 | 1853 | ||
NM_001270774.1 | NP_001257703.1 | 2 | 1853 | |||
NM_001270779.1 | NP_001257708.1 | 2 | 1853 | |||
NM_019268.3 | NP_062141.3 | 2 | 1853 | |||
XM_006239660.2 | XP_006239722.1 | 3 | 2370 | |||
XM_008764432.1 | XP_008762654.1 | 3 | 2017 | |||
XM_008764434.1 | XP_008762656.1 | 3 | 2016 | |||
XM_008764436.1 | XP_008762658.1 | 3 | 2016 | |||
XM_008764438.1 | XP_008762660.1 | 3 | 2016 | |||
GenBank mRNA | AY245282.1 | - | 2 | 107 | ||
U04937.1 | - | 2 | 107 | |||
U04939.1 | - | 2 | 107 | |||
U04940.1 | - | 2 | 107 | |||
X68191.1 | - | 2 | 1882 | |||
X68812.1 | - | 2 | 1933 | |||
X68813.1 | - | 2 | 1944 |