Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FSCN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003088.3 | NP_003079.1 | 3 | 1195 | ||
GenBank mRNA | AK095254.1 | - | 4 | 1129 | ||
AK095830.1 | - | 2 | 616 | |||
AK316607.1 | - | 3 | 1190 | |||
BC000521.2 | - | 3 | 1086 | 2961047 | ||
BC006304.2 | - | 3 | 1177 | 4100996 | ||
BC007539.1 | - | 3 | 1086 | |||
BC007643.1 | - | 3 | 819 | |||
BC007948.2 | - | 3 | 1174 | 4299524 | ||
BC007988.2 | - | 3 | 1086 | 2961047 | ||
BT006636.1 | - | 3 | 1063 | |||
U03057.1 | - | 3 | 1174 | |||
U09873.1 | - | 3 | 1108 |