Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRNP70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301069.1 | NP_001287998.1 | 7, 8 | 983 | ||
NM_003089.5 | NP_003080.2 | 7, 8 | 983 | |||
XM_011527240.2 | XP_011525542.2 | 2, 3 | 202 | |||
GenBank mRNA | AK293120.1 | - | 7, 8 | 702 | ||
AL560281.3 | - | 7, 8 | 671 | |||
BC001315.1 | - | 7, 8 | 656 | 3454479 | ||
M22636.1 | - | 7, 8 | 647 | |||
X06812.1 | - | 5, 6 | 331 | |||
X84841.1 | - | 6, 7 | 476 |