Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apbb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080478.1 | NP_536726.1 | 1 | 408 | ||
XM_006229935.1 | XP_006229997.1 | 2 | 777 | |||
XM_006229936.2 | XP_006229998.1 | 2 | 613 | |||
XM_006229938.2 | XP_006230000.1 | 2 | 952 | |||
GenBank mRNA | AF333983.1 | - | 1 | 408 |