Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004596.4 | NP_004587.1 | 6 | 1359 | ||
GenBank mRNA | AK293140.1 | - | 5 | 755 | ||
AK313182.1 | - | 6 | 972 | |||
BC000405.2 | - | 6 | 1339 | 2822987 | ||
BC008290.1 | - | 6 | 923 | 3677365 | ||
X06347.1 | - | 6 | 929 |