Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004596.4 | NP_004587.1 | 2, 3 | 802 | ||
GenBank mRNA | AK313182.1 | - | 2, 3 | 415 | ||
BC000405.2 | - | 2, 3 | 782 | 2822987 | ||
BC008290.1 | - | 2, 3 | 366 | 3677365 | ||
X06347.1 | - | 2, 3 | 372 |