Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sema6c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017308.1 | NP_059004.1 | 12 | 1433 | ||
XM_006232858.2 | XP_006232920.1 | 15 | 1845 | |||
XM_006232861.2 | XP_006232923.1 | 13 | 1707 | |||
XM_006232869.2 | XP_006232931.1 | 11 | 1354 | |||
XM_008761286.1 | XP_008759508.1 | 13 | 5111 | |||
XM_008761287.1 | XP_008759509.1 | 13 | 4458 | |||
XM_008761288.1 | XP_008759510.1 | 13 | 3384 | |||
XM_008761289.1 | XP_008759511.1 | 13 | 4963 | |||
XM_008761290.1 | XP_008759512.1 | 13 | 4210 | |||
XM_008761291.1 | XP_008759513.1 | 13 | 4541 | |||
XM_008761292.1 | XP_008759514.1 | 13 | 3640 | |||
XM_008761293.1 | XP_008759515.1 | 14 | 1797 | |||
XM_008761294.1 | XP_008759516.1 | 12 | 5269 | |||
GenBank mRNA | AB000817.2 | - | 12 | 1449 | ||
AB014074.1 | - | 12 | 1449 | |||
AF363971.1 | - | 11 | 1313 |