Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnmb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019273.1 | NP_062146.1 | 606 | |||
XM_006246089.2 | XP_006246151.1 | 2112 | ||||
XM_008767562.1 | XP_008765784.1 | 3064 | ||||
XM_008767563.1 | XP_008765785.1 | 2931 | ||||
XM_008767564.1 | XP_008765786.1 | 3064 | ||||
GenBank mRNA | AB010963.1 | - | 315 | |||
AB050745.1 | - | 293 | ||||
AF020712.1 | - | 606 | ||||
FJ154955.1 | - | 295 | ||||
U40602.1 | - | 293 | ||||
U54498.1 | - | 344 |