Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sema3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017310.1 | NP_059006.1 | 10 | 1057 | ||
XM_006235989.2 | XP_006236051.1 | 12 | 1444 | |||
XM_006235990.2 | XP_006236052.1 | 11 | 1297 | |||
XM_008762702.1 | XP_008760924.1 | 12 | 1306 | |||
GenBank mRNA | X95286.1 | - | 10 | 1057 |