Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105729.3 | NP_001099199.2 | 3 | 283 | ||
XM_006228677.2 | XP_006228739.1 | 3 | 320 | |||
XM_006228678.2 | XP_006228740.1 | 3 | 317 | |||
XM_006228679.2 | XP_006228741.1 | 3 | 317 | |||
XM_006228680.2 | XP_006228742.1 | 3 | 322 | |||
XM_006228681.2 | XP_006228743.1 | 3 | 318 | |||
XM_006228682.2 | XP_006228744.1 | 3 | 324 | |||
XM_006228684.2 | XP_006228746.1 | 3 | 1019 | |||
XM_006228685.1 | XP_006228747.1 | 3 | 1144 | |||
XM_008759180.1 | XP_008757402.1 | 3 | 306 | |||
GenBank mRNA | BC159415.1 | - | 3 | 211 | 8368092 | |
CO396551.1 | - | 3 | 283 | |||
X66022.1 | - | 3 | 1144 |