Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp4a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175837.1 | NP_787031.1 | 4, 5 | 538 | ||
XM_006238712.2 | XP_006238774.1 | 4, 5 | 550 | |||
GenBank mRNA | BC089761.1 | - | 4, 5 | 516 | 7324500 | |
X07259.1 | - | 4, 5 | 508 |