Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRPC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003093.2 | NP_003084.1 | 3 | 550 | ||
NR_029472.1 | 2 | 507 | ||||
GenBank mRNA | AK311955.1 | - | 3 | 130 | ||
BC121082.1 | - | 3 | 119 | 40119776 | ||
BC121083.1 | - | 3 | 119 | 40119777 | ||
BU153661.1 | - | 2 | 115 | |||
BU508721.1 | - | 2 | 76 | |||
CD109745.1 | - | 3 | 567 | |||
M18465.1 | - | 1 | 28 | |||
X12517.1 | - | 3 | 115 |