Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnj3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031610.3 | NP_113798.1 | 2 | 819 | ||
GenBank mRNA | L25264.1 | - | 2 | 819 | ||
U01071.1 | - | 2 | 792 | |||
U01141.1 | - | 2 | 833 | |||
U09243.1 | - | 947 | ||||
U72410.1 | - | 2 | 333 |