Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CALM2 SIRNA Assays ›
Gene Symbol:
Calm2
Gene Name:
calmodulin 2
Gene Aliases:
Cam
Chromosome Location:
Chr.6: 11064750 - 11080134 on Build Rnor_6.0
UniGene ID:
Rn.5968
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
s132297
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Calm2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
GenBank mRNA
BC058485.1
-
414
6921326
FQ211494.1
-
414
FQ211970.1
-
410
FQ212357.1
-
411
FQ212640.1
-
410
FQ212691.1
-
410
FQ212789.1
-
414
FQ212869.1
-
414
FQ213028.1
-
410
FQ213029.1
-
408
FQ213091.1
-
410
FQ213115.1
-
416
FQ213200.1
-
413
FQ213216.1
-
410
FQ213310.1
-
303
FQ213366.1
-
409
FQ213496.1
-
413
FQ213520.1
-
414
FQ213526.1
-
410
FQ213581.1
-
410
FQ213598.1
-
410
FQ213695.1
-
410
FQ213703.1
-
410
FQ213760.1
-
410
FQ213762.1
-
415
FQ213913.1
-
410
FQ213941.1
-
414
FQ213981.1
-
411
FQ213985.1
-
304
FQ213999.1
-
409
FQ214059.1
-
414
FQ214181.1
-
414
FQ214262.1
-
414
FQ214491.1
-
410
FQ214500.1
-
410
FQ214519.1
-
410
FQ214525.1
-
410
FQ214540.1
-
414
FQ215159.1
-
414
FQ215317.1
-
410
FQ215600.1
-
414
FQ216397.1
-
410
FQ216471.1
-
411
FQ218091.1
-
410
FQ220250.1
-
410
FQ220325.1
-
410
FQ220698.1
-
414
FQ220779.1
-
410
FQ222184.1
-
413
FQ223357.1
-
414
FQ225272.1
-
410
FQ226994.1
-
414
FQ229228.1
-
410
FQ229580.1
-
414
FQ229619.1
-
410
FQ229790.1
-
410
FQ229905.1
-
409
FQ231022.1
-
414
FQ231702.1
-
414
FQ233337.1
-
413
FQ234377.1
-
414
FQ235266.1
-
411
M17069.1
-
412
M19312.1
-
393
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Human: NM_001743, BT009916, AK315694, D45887, M19311, and others.
Panther Classification:
Molecular Function -
calmodulin-related
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-8tsm6:80/100.66.77.47:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline