Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291916.1 | NP_001278845.1 | 3 | 285 | ||
NM_006938.3 | NP_008869.1 | 3 | 408 | |||
GenBank mRNA | AB451227.1 | - | 1 | 247 | ||
AB451350.1 | - | 1 | 247 | |||
AJ577268.1 | - | 3 | 340 | |||
BC001721.1 | - | 3 | 369 | 3355716 | ||
BC072427.1 | - | 3 | 295 | 6464946 | ||
BU565530.1 | - | 3 | 410 | |||
CN354758.1 | - | 3 | 221 | |||
CR542239.1 | - | 1 | 247 | |||
HY043971.1 | - | 3 | 406 | |||
J03798.1 | - | 3 | 397 |