Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017337.1 | NP_059033.1 | 3 | 1196 | ||
XM_008763422.1 | XP_008761644.1 | 3 | 1264 | |||
GenBank mRNA | U38179.1 | - | 3 | 1196 |