Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myl12a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135017.1 | NP_001128489.1 | 3 | 456 | ||
XM_006245667.2 | XP_006245729.1 | 3 | 610 | |||
XM_006245668.2 | XP_006245730.1 | 3 | 551 | |||
GenBank mRNA | EV767772.1 | - | 486 | |||
FQ212603.1 | - | 1 | 160 | |||
FQ215121.1 | - | 3 | 411 | |||
FQ215776.1 | - | 600 | ||||
FQ217399.1 | - | 3 | 502 | |||
FQ217558.1 | - | 3 | 386 | |||
FQ220614.1 | - | 442 | ||||
FQ223132.1 | - | 3 | 420 | |||
FQ228530.1 | - | 3 | 412 | |||
FQ229140.1 | - | 3 | 665 | |||
X05566.1 | - | 3 | 398 | |||
Z32519.1 | - | 3 | 398 |
Gene Symbol: Myl12b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017343.2 | NP_059039.2 | 3 | 429 | ||
GenBank mRNA | BC060577.1 | - | 3 | 428 | 6889406 | |
EV769828.1 | - | 3 | 416 | |||
FQ215510.1 | - | 3 | 415 | |||
FQ229195.1 | - | 3 | 415 | |||
X52840.1 | - | 2 | 346 |