Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ntm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017354.1 | NP_059050.1 | 5 | 1256 | ||
XM_006242749.1 | XP_006242811.1 | 5 | 1574 | |||
XM_008766065.1 | XP_008764287.1 | 5 | 1574 | |||
GenBank mRNA | U16845.1 | - | 5 | 1256 |