Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRPD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278656.1 | NP_001265585.1 | 2 | 499 | ||
NM_004175.4 | NP_004166.1 | 2 | 513 | |||
NR_103819.1 | 2 | 513 | ||||
GenBank mRNA | AB451249.1 | - | 1 | 73 | ||
AB451373.1 | - | 1 | 73 | |||
AK296173.1 | - | 2 | 156 | |||
BC000457.2 | - | 2 | 169 | 2822042 | ||
BC003150.1 | - | 2 | 169 | 2822042 | ||
BC034447.1 | - | 2 | 196 | |||
BF344644.1 | - | 2 | 508 | |||
BM928709.1 | - | 2 | 461 | |||
BP202359.1 | - | 2 | 163 | |||
BY993733.1 | - | 2 | 185 | |||
CN286602.1 | - | 2 | 153 | |||
CR456583.1 | - | 2 | 189 | |||
U15009.1 | - | 2 | 160 |