Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRPE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304464.1 | NP_001291393.1 | 3 | 238 | ||
NM_003094.3 | NP_003085.1 | 3 | 194 | |||
NR_130746.1 | 2 | 197 | ||||
GenBank mRNA | AI263891.1 | - | 2 | 173 | ||
AK307255.1 | - | 3 | 224 | |||
AK312130.1 | - | 3 | 171 | |||
BC002639.2 | - | 3 | 163 | 3609230 | ||
BC072433.1 | - | 3 | 142 | |||
BC090951.1 | - | 3 | 167 | 5204266 | ||
CN272809.1 | - | 2 | 122 | |||
DW441956.1 | - | 3 | 212 | |||
M15919.1 | - | 3 | 87 | |||
M37716.1 | - | 3 | 164 | |||
X12466.1 | - | 3 | 164 |