Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pbsn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019125.1 | NP_061998.1 | 239 | |||
GenBank mRNA | BC062035.1 | - | 217 | 5623889 | ||
M27156.1 | - | 239 |
Gene Symbol: LOC103689993 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773239.1 | XP_008771461.1 | 2 | 339 |