Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crebbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133381.3 | NP_596872.3 | 19, 20 | 3718 | ||
XM_008767524.1 | XP_008765746.1 | 18, 19 | 3434 | |||
GenBank mRNA | AY462245.1 | - | 19, 20 | 3718 |