Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cfd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077642.1 | NP_001071110.1 | 4 | 523 | ||
GenBank mRNA | M92059.1 | - | 4 | 520 | ||
S73894.1 | - | 4 | 523 |