Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019315.2 | NP_062188.2 | 1 | 1189 | ||
GenBank mRNA | AF284345.1 | - | 1 | 886 | ||
AF292389.1 | - | 1 | 1189 | |||
U69884.1 | - | 1 | 893 |