Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Llgl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006246518.2 | XP_006246580.1 | 6 | 786 | ||
XM_006246519.2 | XP_006246581.1 | 6 | 786 | |||
GenBank mRNA | AF356187.1 | - | 6 | 631 |