Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcpt10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017146.1 | NP_058842.1 | 252 | |||
GenBank mRNA | U67913.1 | - | 49 | |||
X68657.1 | - | 252 |
Gene Symbol: LOC691603 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751469.3 | XP_003751517.1 | 3 | 312 |
Gene Symbol: Mcpt8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021598.2 | NP_067609.1 | 251 | |||
GenBank mRNA | FQ229669.1 | - | 270 | |||
U67911.1 | - | 252 |
Gene Symbol: Mcpt8l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_573781.6 | XP_573781.1 | 3 | 314 |