Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr4a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019328.3 | NP_062201.2 | 6 | 1591 | ||
XM_006234201.2 | XP_006234263.1 | 7 | 1809 | |||
XM_008761861.1 | XP_008760083.1 | 6 | 1725 | |||
GenBank mRNA | DQ092632.1 | - | 6 | 1472 | ||
DQ092633.1 | - | 6 | 1358 | |||
L08595.1 | - | 6 | 1588 | |||
U01146.1 | - | 6 | 1522 | |||
U72345.1 | - | 7 | 1815 |